Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
5 L
25 L
Packaging Type:
View all
Glass Bottle
Catalog number FSA233360050
Price (MYR)
244.00
Add to cart
Quantity:
5 mL
Packaging Type:
Glass Bottle
Request bulk or custom format
Price (MYR)
244.00
Add to cart
Chemical Identifiers
CAS1330-78-5
SpecificationsSpecification SheetSpecification Sheet
GC63 to 85 % (sum of Tritolyl phosphate-isomers)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.5550 to 1.5600 (20°C, 589 nm)
Appearance (Form)Liquid