Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
5 L
25 L
Packaging Type:
View all
Glass Bottle
Catalog number FSA233360010
Price (MYR)
576.00
Add to cart
Quantity:
1 L
Packaging Type:
Glass Bottle
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Chemical Identifiers
CAS98-06-6
IUPAC Nametert-butylbenzene
Molecular FormulaC10H14
InChI KeyYTZKOQUCBOVLHL-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4910-1.4940 @ 20?C