Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
Iron(II) sulfide, fused sticks
CAS: 1317-37-9 | FeS | 87.91 g/mol
Have Questions?
Catalog number FSA233300010
Price (MYR)
552.00
Add to cart
Request bulk or custom format
Price (MYR)
552.00
Add to cart
Specifications
Chemical Name or MaterialIron(II) sulfide
Melting Point1195.0°C
Name Notec.p., 60-67% Fe
CAS1317-37-9
ColorBlack-Gray