2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals
CAS: 610-14-0 | C7H4ClNO3 | 185.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA232310050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5720-07-0
IUPAC Name(4-methoxyphenyl)boronic acid
Molecular FormulaC7H9BO3
InChI KeyVOAAEKKFGLPLLU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.0%
Assay (HPLC)≥97.0%
Identification (FTIR)Conforms
Proton NMRConforms to structure
Appearance (Color)White to pale cream to pale brown