Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Glass Bottle
Catalog number FSA232110025
Price (MYR)
719.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
719.00
Add to cart
Chemical Identifiers
CAS56777-24-3
IUPAC Namebenzyl 2-hydroxypropanoate
Molecular FormulaC10H12O3
InChI KeyZYTLPUIDJRKAAM-UHFFFAOYNA-N
SMILESCC(O)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.5100-1.5150 @ 20?C
FormLiquid
Optical Rotation-16.8 ± 0.5? (C=3 in methanol)
For analysis
RUO – Research Use Only