n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA232100010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1758-73-2
IUPAC Namecarbamimidoylsulfinic acid
Molecular FormulaCH4N2O2S
InChI KeyFYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILESNC(=N)S(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Water=<0.5 %
Appearance (Color)White to light yellow
Titration with NaOH>=98.5 %
Infrared spectrumConforms
View more
For HPLC
RUO – Research Use Only