Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Thermo Scientific Chemicals
Dodecyl sulfate, sodium salt, 99%, for biochemistry
CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Catalog number FSA230421000
Price (MYR)
539.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Chemical Identifiers
CAS467-62-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrumComplies
Appearance (Color)Purple
For biochemistry
RUO – Research Use Only