D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals
D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
25 g
Packaging:
View all
Glass bottle
Catalog number FSA230090050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1313-27-5
IUPAC Nametrioxomolybdenum
Molecular FormulaMoO3
InChI KeyJKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILESO=[Mo](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.05%
Appearance (Color)White to cream to yellow to green