γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA229905000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12030-24-9
IUPAC Namediindium(3+) trisulfanediide
Molecular FormulaIn2S3
InChI KeySIXIBASSFIFHDK-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[In+3].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Chelation/Complexation reagent
RUO – Research Use Only