γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
Packaging:
Glass bottle
Catalog number FSA229900050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14284-92-5
IUPAC Namerhodium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21O6Rh
InChI KeyDGOINFUDFBWCMX-LNTINUHCSA-K
SMILES[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=24.7 % (Rh)
Appearance (Color)Yellow to light orange
Infrared spectrumConforms
Appearance (Form)Powder
Chelation/Complexation reagent
RUO – Research Use Only