Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
250 g
Packaging:
Glass bottle
Catalog number FSA228940500
Price (MYR)
532.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
532.00
Add to cart
Chemical Identifiers
CAS16925-25-0
Molecular FormulaF6NaSb
InChI KeyZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol)258.74
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 %