Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Creatinine, 98%, Thermo Scientific Chemicals
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA228940250
Price (MYR)
361.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
361.00
Add to cart
Chemical Identifiers
CAS23598-72-3
IUPAC Name3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular FormulaC11H8ClNO3
InChI KeyUVEPOHNXGXVOJE-UHFFFAOYSA-N
SMILESCC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow or cream to pale brown
FormPowder
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Assay (GC)≥96.0%