5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
5-Bromo-2'-deoxyuridine, 99+%
CAS: 59-14-3 | C9H11BrN2O5 | 307.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
500 mg
1 g
2.5 g
Packaging:
Glass bottle
Catalog number FSA228590100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS76195-82-9
IUPAC Name(2,4,6-trimethylphenyl)hydrazine hydrochloride
Molecular FormulaC9H15ClN2
InChI KeyXFGPCULNJGZRTH-UHFFFAOYSA-N
SMILESCl.CC1=CC(C)=C(NN)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to cream to orange to pale brown