D-Cycloserine, 98%
D-Cycloserine, 98%
D-Cycloserine, 98%
Thermo Scientific Chemicals
D-Cycloserine, 98%
D-Cycloserine, CAS # 68-41-7, is a cyclic amino acid analog of D-alanine, originally isolated from Streptomyces species. It is synthetically formulated and exhibits antibiotic properties through its inhibition of bacterial cell wall synthesis.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA228480010
Price (MYR)
563.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
563.00
Add to cart
Chemical Identifiers
CAS7789-69-7
IUPAC Namepentabromo-λ⁵-phosphane
Molecular FormulaBr5P
InChI KeyQRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILESBrP(Br)(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormCrystalline powder and/or Lumps
Assay (Titration ex Bromide)>94.0 to <106.0%
General description
  • D-Cycloserine was originally isolated from Streptomyces orchidaceus and is currently formulated as an oral antibiotic
  • It is incompatible with strong oxidizing agents but stable under basic conditions, with the greatest stability at pH = 11.5
  • D-Cycloserine inhibits L-alanine racemase and D-alanine:D-alanine ligase, 2 enzymes responsible for bacterial cell wall synthesis
  • When ingested, D-Cycloserine traverses the blood-brain barrier, potentially causing neurological side effects such as, dizziness, nausea, depression, tremors, etc.
Applications
  • D-Cycloserine is a broad-spectrum antibiotic used as a second line agent for treatment of drug-resistant tuberculosis, always in combination with other antituberculosis agents
  • It is an excitatory amino acid and partial agonist of the NMDA receptor in the central nervous system (CNS), thus playing a potential role in the amelioration of neuropathic pain
  • D-Cycloserine has also been shown to play a role in neural connectivity, and is therefore used to study treatment applications in anxiety disorders and schizophrenia
RUO – Research Use Only