D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Camphor, 97%, Thermo Scientific Chemicals
CAS: 464-49-3 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA227841000
Price (MYR)
312.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
312.00
Add to cart
Chemical Identifiers
CAS110-99-6
IUPAC Name2-(carboxymethoxy)acetic acid
Molecular FormulaC4H6O5
InChI KeyQEVGZEDELICMKH-UHFFFAOYSA-N
SMILESOC(=O)COCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Melting point141°C to 145°C
Appearance (Color)White to light beige