(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
(+)-5-Fluoro-2'-deoxyuridine, 99+%
CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number FSA227601000
Price (MYR)
965.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
965.00
Add to cart
Chemical Identifiers
CAS2893-78-9
IUPAC Namesodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular FormulaC3Cl2N3NaO3
InChI KeyMSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (on dried substance)
Appearance (Color)White
FormGranules or granular powder or crystals
Identification (FTIR)Conforms
Loss on Drying≤5.0%