Auramine O, pure, biological stain
Auramine O, pure, biological stain
Auramine O, pure, biological stain
Auramine O, pure, biological stain
Thermo Scientific Chemicals
Auramine O, pure, biological stain
CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol
Have Questions?
Catalog NumberQuantity
FSA22752050050 g
Catalog number FSA227520500
Price (MYR)
942.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
942.00
Add to cart
Chemical Identifiers
CAS1460-57-7
IUPAC Name(1R,2R)-cyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-PHDIDXHHSA-N
SMILESO[C@@H]1CCCC[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point100°C to 104°C
GC>=97.5 %
Appearance (Color)Off-white
Appearance (Form)Crystalline powder or crystals