β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin hydrate, 99%, Thermo Scientific Chemicals
CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA227280250
Price (MYR)
344.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
344.00
Add to cart
Chemical Identifiers
CAS7631-95-0
IUPAC Namedisodium dioxomolybdenumbis(olate)
Molecular FormulaMoNa2O4
InChI KeyTVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofANa₂MoO4: ≥99.2% (≥46.2% Mo Calculated)
FormPowder
Chelation/Complexation reagent
RUO – Research Use Only