5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminouracil, 98%, Thermo Scientific Chemicals
CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA226820250
Price (MYR)
982.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
982.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.