Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA226810010
Price (MYR)
540.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Chemical Identifiers
CAS96-41-3
IUPAC Namecyclopentanol
Molecular FormulaC5H10O
InChI KeyXCIXKGXIYUWCLL-UHFFFAOYSA-N
SMILESOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4515-1.4545 @ 20?C
FormLiquid
Assay (GC)≥98.5%