Thymidine, 99+%
Thymidine, 99+%
Thymidine, 99+%
Thermo Scientific Chemicals
Thymidine, 99+%
CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA226740250
Price (MYR)
1,469.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,469.00
Add to cart
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
Heavy metals (as Pb)=<5 ppm
Potassium (K)=<0.005 %
Sulfate (SO4)=<0.002 %