2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA226720050
Price (MYR)
1,440.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,440.00
Add to cart
Chemical Identifiers
CAS5447-02-9
IUPAC Name3,4-bis(benzyloxy)benzaldehyde
Molecular FormulaC21H18O3
InChI KeyXDDLXZHBWVFPRG-UHFFFAOYSA-N
SMILESO=CC1=CC=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point89°C to 93°C
GC>=98.5 %
Appearance (Color)White to yellow to light beige