2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA226720010
Price (MYR)
428.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
428.00
Add to cart
Chemical Identifiers
CAS594-56-9
IUPAC Name2,3,3-trimethylbut-1-ene
Molecular FormulaC7H14
InChI KeyAUYRUAVCWOAHQN-UHFFFAOYSA-N
SMILESCC(=C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.0 %
Refractive index1.4015 to 1.4035 (20°C, 589 nm)
Appearance (Form)Liquid