Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aristolochic acid I, 96%, Thermo Scientific Chemicals
CAS: 313-67-7 | C17H11NO7 | 341.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA226091000
Price (MYR)
1,257.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,257.00
Add to cart
Chemical Identifiers
CAS349-88-2
IUPAC Name4-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyBFXHJFKKRGVUMU-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Titration Argentometric>=97.5 % (ex Cl)
Infrared spectrumConforms
Appearance (Form)Low melting crystalline mass