hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA223620010
Price (MYR)
1,908.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,908.00
Add to cart
Chemical Identifiers
CAS124-09-4
IUPAC Namehexane-1,6-diamine
Molecular FormulaC6H16N2
InChI KeyNAQMVNRVTILPCV-UHFFFAOYSA-N
SMILESNCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Strength59 to 61 wt%
Color scale=<20 APHA