Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA223310010
Price (MYR)
3,074.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,074.00
Add to cart
Chemical Identifiers
CAS67-21-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to cream
Oxidation reagent for analysis
RUO – Research Use Only