Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Thermo Scientific Chemicals
Sodium hypophosphite monohydrate, for analysis
CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA222792500250 gPlastic bottle
Catalog number FSA222792500
Price (MYR)
595.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Specifications
Chemical Name or MaterialSodium hypophosphite monohydrate
Melting Point90.0°C
CAS10039-56-2
Quantity250 g
Molecular FormulaH4NaO3P