Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Thermo Scientific Chemicals
Sodium hypophosphite monohydrate, for analysis
CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA222791000100 gPlastic bottle
Catalog number FSA222791000
Price (MYR)
364.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
364.00
Add to cart
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.5 %
Appearance (Color)Clear colorless
Water=<0.005 %