Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number FSA222165000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 250 nm A: =<0.05
Ethanol=<0.01 %
Infrared spectrumConforms
Titrable acid0.0004 meq/g
UVneat
View more
For UV/VIS spectroscopy
RUO – Research Use Only