Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA222072500
Price (MYR)
231.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
231.00
Add to cart
Chemical Identifiers
CAS111-65-9
IUPAC Nameoctane
Molecular FormulaC8H18
InChI KeyTVMXDCGIABBOFY-UHFFFAOYSA-N
SMILESCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Density(15°C) 0.706 to 0.710 kg/l
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid