Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
CAS: 5329-14-6 | H3NO3S | 97.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA222070025
Price (MYR)
811.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
811.00
Add to cart
Specifications
Chemical Name or MaterialSulfamic acid
ColorWhite
CAS5329-14-6
Melting Point200.0°C to 208.0°C
Health Hazard 1GHS Signal Word: Warning