Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA221802500
Price (MYR)
424.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% ZnO min
Impurity contentAlkalinity P.T.
Impurity contentFe 0.001%
Impurity contentInsoluble in dilute sulfuric acid 0.01%
Impurity contentK 0.01%
View more
For analysis
RUO – Research Use Only