Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
CAS: 87749-50-6 | C16H36FN·3H2O | 315.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA221080500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS629-59-4
IUPAC Nametetradecane
Molecular FormulaC14H30
InChI KeyBGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
C-X bond formation reagent
RUO – Research Use Only