Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number FSA220890010
Price (MYR)
756.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
756.00
Add to cart
Chemical Identifiers
CAS206658-83-5
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)123.0-130.0?C
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)Dark cream to cream to pale brown
Water Content (Karl Fischer Titration)4-11.11% (0.5-1.5 waters)
Biological buffer
RUO – Research Use Only