Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromoform, 99+%, stabilized, Thermo Scientific Chemicals
CAS: 75-25-2 | CHBr3 | 252.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA220695000
Price (MYR)
4,347.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,347.00
Add to cart
Chemical Identifiers
CAS867-13-0
IUPAC Nameethyl 2-(diethoxyphosphoryl)acetate
Molecular FormulaC8H17O5P
InChI KeyGGUBFICZYGKNTD-UHFFFAOYSA-N
SMILESCCOC(=O)CP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Acid value=<2 mg KOH/g
Infrared spectrumConforms
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
GC>=96.0 %