1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA220620050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Butanethiol
Name Note99+%
CAS109-79-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger