1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
Packaging:
Glass bottle
Catalog number FSA220620010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS367-86-2
IUPAC Name1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Molecular FormulaC7H3F4NO2
InChI KeyHLDFCCHSOZWKAA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4595-1.4645 @ 20?C
Appearance (Color)Clear yellow to orange