Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA220341000
Price (MYR)
413.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
413.00
Add to cart
Chemical Identifiers
CAS114615-82-6
IUPAC Nametetrapropylazanium tetrahydrate ruthenium
Molecular FormulaC12H36NO4Ru
InChI KeyHUCLFLGLPCVDMZ-UHFFFAOYSA-N
SMILESO.O.O.O.[Ru].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Assay (unspecified)>97.5% (based on elemental Ru)
Formpowder
Biological buffer for analysis
RUO – Research Use Only