Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, Thermo Scientific Chemicals
CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number FSA220320010
Price (MYR)
502.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Chemical Identifiers
CAS122965-43-9
IUPAC Name7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC16H18ClN3S
InChI KeyCXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<22 %
Appearance (Color)Green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with TiCl3>=70 %