sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA220290010
Price (MYR)
443.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
443.00
Add to cart
Chemical Identifiers
CAS37348-16-6
SpecificationsSpecification SheetSpecification Sheet
Formfuming liquid
Appearance (Color)Pale to dark yellow
Assay (Titration ex Bromide)31 - 34.5% w/w HBr