Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Catalog number FSA220050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight76.12
GradeAnalysis
Unit Size
For analysis
RUO – Research Use Only