Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA219682500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1629-60-3
IUPAC Namehex-1-en-3-one
Molecular FormulaC6H10O
InChI KeyJTHNLKXLWOXOQK-UHFFFAOYSA-N
SMILESCCCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥90.0%
Refractive Index1.4240-1.4310 @ 20?C
Solution Test10% w/v in diethyl ether: clear solution