Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA219680500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS286-20-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4500 to 1.4540 (20°C, 589 nm)
GC>=97.5 %