2-Ethylimidazole, 99%, Thermo Scientific Chemicals
2-Ethylimidazole, 99%, Thermo Scientific Chemicals
2-Ethylimidazole, 99%, Thermo Scientific Chemicals
2-Ethylimidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethylimidazole, 99%, Thermo Scientific Chemicals
CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA219281000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1072944-98-9
IUPAC Nametert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate
Molecular FormulaC15H24BNO4
InChI KeyDZQURIUJAGTAFW-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1C=CC=C1B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White
Assay from Supplier's CofA≥96.0%