Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA219221000
Price (MYR)
668.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
668.00
Add to cart
Chemical Identifiers
CAS2802-68-8
IUPAC Namebis(methanol); trifluoroborane
Molecular FormulaC2H8BF3O2
InChI KeyMPRMFRXCVMCOMX-UHFFFAOYSA-N
SMILESCO.CO.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay50 to 53 wt% (BF3)
Appearance (Form)Liquid
Density(20°C) 1.210 to 1.235 kg/l
Appearance (Color)Clear colorless to yellow