Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
CAS: 458-37-7 | C21H20O6 | 368.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA218580500
Price (MYR)
1,718.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,718.00
Add to cart
Chemical Identifiers
CAS104663-56-1
IUPAC Namemanganese(3+) phosphate
Molecular FormulaMnO4P
InChI KeyBECVLEVEVXAFSH-UHFFFAOYSA-K
SMILES[Mn+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale grey to pale brown or green
FormPowder or crystalline powder
Complexometric Titration(Manganese, Mn): 36.1-37.2% (dry wt. basis)
Complexometric Titration≥98.5 to ≤101.5% (dry wt. basis)
Loss on Drying≤20% (200°C/constant wt.)