Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%, Thermo Scientific Chemicals
CAS: 458-37-7 | C21H20O6 | 368.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA218580100
Price (MYR)
436.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS112-72-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes or
Appearance (Form)pellets and/or chunks
Infrared spectrumConforms
Melting point37°C to 41°C
GC>=98.5 %