Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA218210010
Price (MYR)
2,510.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,510.00
Add to cart
Specifications
Chemical Name or MaterialPhenyl chloroformate
CAS1885-14-9
Health Hazard 1GHS Signal Word: Danger
Melting Point-28.0°C
Density1.2440g/mL
View more
Coupling reagent
RUO – Research Use Only