Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA217421000100 mLAcroSeal™ Glass Bottle
Catalog number FSA217421000
Price (MYR)
4,239.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
4,239.00
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Total Metal Impurities≤0.5%
pH6.5-7.5 (suspension 5% w/w)
Particle SizeOver size, air jet sieve alpine 60 micrometer: ≤10% (w/w)
View more
For NMR
RUO – Research Use Only