Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
FSA2173400505 mL
Catalog number FSA217340050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl sulfoxide-d6
Density1.1900g/mL
Deuterium (D)99.95% min.
Flash Point88°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only